Skip to Content
Merck
Ciclopirox Related Compound C certified reference material, pharmaceutical secondary standard

Ciclopirox Related Compound C

Ciclopirox Related Compound C certified reference material, pharmaceutical secondary standard
Synonym(s):
6-Cyclohexyl-4-methylpyridin-2(1H)-one
Empirical Formula (Hill Notation):
C12H17NO
CAS Number:
Molecular Weight:
191.27

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade