![Chloramphenicol sodium succinate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/240/006/3b70d07b-799e-4905-9816-1cd8af4207f6/640/3b70d07b-799e-4905-9816-1cd8af4207f6.png)
Chloramphenicol sodium succinate
![Chloramphenicol sodium succinate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/240/006/3b70d07b-799e-4905-9816-1cd8af4207f6/640/3b70d07b-799e-4905-9816-1cd8af4207f6.png)
Synonym(s):
Chloramphenicol succinate sodium salt, Chloramphenicol α-succinate
Linear Formula:
C15H15Cl2N2O8Na
CAS Number:
Molecular Weight:
445.18
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2