![Chloramphenicol Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/257/425/5b518d62-a18a-46be-959b-459222e72dbf/640/5b518d62-a18a-46be-959b-459222e72dbf.jpg)
Chloramphenicol
![Chloramphenicol Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/257/425/5b518d62-a18a-46be-959b-459222e72dbf/640/5b518d62-a18a-46be-959b-459222e72dbf.jpg)
Synonym(s):
D-(-)-threo-2,2-Dichloro-N-[β-hydroxy-α−(hydroxymethyl)-β-(4-nitrophenyl)ethyl]acetamide, D-(-)-threo-2-Dichloroacetamido-1-(4-nitrophenyl)-1,3-propanediol, D-threo-2,2-Dichloro-N-[β-hydroxy-α-(hydroxymethyl)-4-nitrophenethyl]acetamide, Chloramphenicol
EC Number:
Products
Product Category
Brand
Physical Form
Purity
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
Optical Isomer
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2