Chelidonic acid
Synonym(s):
4-Oxo-4H-pyran-2,6-dicarboxylic acid
Empirical Formula (Hill Notation):
C7H4O6
CAS Number:
Molecular Weight:
184.10
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing