
CGP-42112A

Synonym(s):
Nα-Nicotinoyl-Tyr-(Nα-Cbz-Arg)-Lys-His-Pro-Ile
Empirical Formula (Hill Notation):
C52H69N13O11
CAS Number:
Molecular Weight:
1052.18
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing