![Cefdinir Related Compound B certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/131/413/5f5a3cab-3274-4473-be79-812d0468cfa7/640/5f5a3cab-3274-4473-be79-812d0468cfa7.jpg)
Cefdinir Related Compound B
![Cefdinir Related Compound B certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/131/413/5f5a3cab-3274-4473-be79-812d0468cfa7/640/5f5a3cab-3274-4473-be79-812d0468cfa7.jpg)
Synonym(s):
(6R,7R)-7-[2-(2-Amino-4-thiazolyl)acetamido]-8-oxo-3-vinyl-5-thia-1-azabicyclo[4,2,0]oct-2-ene-2-carboxylic acid, (6R-trans)-7-[[(2-Amino-4-thiazolyl)acetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Empirical Formula (Hill Notation):
C14H14N4O4S2
CAS Number:
Molecular Weight:
366.42
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2