Skip to Content
Merck
CdTe core-type quantum dots COOH functionalized, fluorescence λem 770 nm, powder

CdTe core-type quantum dots

CdTe core-type quantum dots COOH functionalized, fluorescence λem 770 nm, powder
Synonym(s):
Fluorescent nanocrystals, QDs, artificial atoms, CdTe core-type quantum dots
Linear Formula:
CdTe
EC Number:

Products

Product Category
Brand
Physical Form
Quality Segment