
Carvedilol Related Compound B

Synonym(s):
1,1′-{[2-(2-Methoxyphenoxy)ethyl]nitrilo}bis[3-(9H-carbazol-4-yloxy)-propan-2-ol], 3,3′-[2-(2-Methoxyphenoxy)ethylazanediyl]bis[1-(9H-carbazol-4-yloxy)propan-2-ol]
Empirical Formula (Hill Notation):
C39H39N3O6
CAS Number:
Molecular Weight:
645.74
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2