Carpropamid
Synonym(s):
2,2-Dichloro-N-[1-(4-chlorophenyl)ethyl]-1-ethyl-3-methylcyclopropanecarboxamide
Empirical Formula (Hill Notation):
C15H18Cl3NO
CAS Number:
Molecular Weight:
334.67
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing