Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Carbon monoxide in nitrogen
Products
Manufacturer
Physical Form
Format
Quality Segment
Application
Reference Material Type
Special Grade
certified reference material (7)
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
NIST1680B
Carbon monoxide in nitrogen,
NIST
®
SRM
®
1680b, nominal 500 umol/mol
Expand
View Pricing
NIST1679C
Carbon monoxide in nitrogen,
NIST
®
SRM
®
1679c,nominal 100 μmol/mol
Expand
View Pricing
NIST2640A
Carbon monoxide in nitrogen,
NIST
®
SRM
®
2640a, nominal 2% mol/mol
Expand
View Pricing
NIST2740A
Carbon monoxide in nitrogen,
NIST
®
SRM
®
2740a, nominal 10% mol/mol
Expand
View Pricing
NIST2635A
Carbon monoxide in nitrogen,
NIST
®
SRM
®
2635a, nominal 25 umol/mol
Expand
View Pricing
NIST1677C
Carbon monoxide in nitrogen,
NIST SRM
1677c, nominal 10 umol/mol
Expand
View Pricing
NIST1681B
Carbon monoxide in nitrogen,
NIST SRM
1681b, nominal 1000 umol/mol
Expand
View Pricing
NIST2636A
Carbon monoxide in nitrogen,
NIST
®
SRM
®
2636a, nominal 250 umol/mol
Expand
View Pricing
NIST2637A
Carbon monoxide in nitrogen,
NIST
®
SRM
®
2637a, nominal 2500 umol/mol
Expand
View Pricing
NIST2638A
Carbon monoxide in nitrogen,
NIST SRM 2638a, nominal 5000 umol/mol
Expand
View Pricing