![Carbamazepine Impurity F Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/407/098/2961006c-9a3d-4aed-a20e-35b97bfa45c2/640/2961006c-9a3d-4aed-a20e-35b97bfa45c2.png)
Carbamazepine Impurity F
![Carbamazepine Impurity F Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/407/098/2961006c-9a3d-4aed-a20e-35b97bfa45c2/640/2961006c-9a3d-4aed-a20e-35b97bfa45c2.png)
Synonym(s):
Dibenz [b,f]azepine-5-carbonyl chloride
Empirical Formula (Hill Notation):
C15H10ClNO
CAS Number:
Molecular Weight:
255.70
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing