Capric alcohol
Synonym(s):
1-Decanol, n-Decyl alcohol, Alcohol C10
Linear Formula:
CH3(CH2)9OH
CAS Number:
Molecular Weight:
158.28
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing