Skip to Content
Merck
Calcium silicate −200 mesh, 99%

Calcium silicate

Calcium silicate −200 mesh, 99%
Synonym(s):
Calcium metasilicate, Calcium silicon oxide, Silicon calcium oxide
Linear Formula:
CaSiO3
CAS Number:
Molecular Weight:
116.16
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing