
Calcium Ionophore A23187 hemicalcium salt

Synonym(s):
A23187 hemicalcium salt, Antibiotic A 23187 hemicalcium salt, Calcimycin hemicalcium salt, Calimycin hemicalcium salt
Empirical Formula (Hill Notation):
C29H36Ca0.5N3O6
CAS Number:
Molecular Weight:
542.65
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Activity Spectrum
Quality Segment
Mode of Action
Sterility
Showing 1-1 of 1