Skip to Content
Merck
Cadmium acetate dihydrate reagent grade, 98%

Cadmium acetate dihydrate

Cadmium acetate dihydrate reagent grade, 98%
Synonym(s):
Cadmium acetate dihydrate
Linear Formula:
(CH3COO)2Cd · 2H2O
CAS Number:
Molecular Weight:
266.53

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Core
Special Grade
Showing 1-3 of 3