Skip to Content
Merck
Buspirone Related Compound L certified reference material, pharmaceutical secondary standard

Buspirone Related Compound L

Buspirone Related Compound L certified reference material, pharmaceutical secondary standard
Synonym(s):
8-(4-Chlorobutyl)-8-azaspiro[4.5]decane-7,9-dione
Empirical Formula (Hill Notation):
C13H20ClNO2
CAS Number:
Molecular Weight:
257.76

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade