Skip to Content
Merck
Buprenorphine hydrochloride British Pharmacopoeia (BP) Reference Standard

Buprenorphine hydrochloride

Buprenorphine hydrochloride British Pharmacopoeia (BP) Reference Standard
Synonym(s):
[5α,7α(S)]-17-(Cyclopropylmethyl)-α-(1,1-dimethylethyl)4,5-epoxy-18,19-dihydo-3-hydroxy-6-methoxy-α-methyl-6,14-ethenomorphinan-7-methanol hydrochloride
Empirical Formula (Hill Notation):
C29H41NO4 · HCl
CAS Number:
Molecular Weight:
504.10
EC Number:

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade