Skip to Content
Merck
Bromocresol Purple sodium salt indicator grade, Dye content 90 %

Bromocresol Purple sodium salt

Bromocresol Purple sodium salt indicator grade, Dye content 90 %
Synonym(s):
5′,5″-Dibromo-o-cresolsulfonphthalein sodium salt
Empirical Formula (Hill Notation):
C21H15Br2NaO5S
CAS Number:
Molecular Weight:
562.20
EC Number:

Products

Product Category
Color
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing