Skip to Content
Merck
Brinzolamide certified reference material, pharmaceutical secondary standard

Brinzolamide

Brinzolamide certified reference material, pharmaceutical secondary standard
Synonym(s):
(5R)-5-Ethylamino-3-(3-methoxypropyl)-2,2-dioxo-2?6,9-dithia-3-azabicyclo[4.3.0]nona-7,10-diene-8-sulfonamide, (R)- 4-(ethylamino)-3,4-dihydro-2-(3-methoxypropyl)-2H-Thieno[3,2-e]-1,2-thiazine-6-sulfonamide 1,1-dioxide, AL 4862
Empirical Formula (Hill Notation):
C12H21N3O5S3
CAS Number:
Molecular Weight:
383.51

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Special Grade