![Brilliant blue FCF certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/402/366/d27ba2b1-c037-4949-94e4-ea9609405108/640/d27ba2b1-c037-4949-94e4-ea9609405108.png)
Brilliant Blue FCF
![Brilliant blue FCF certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/402/366/d27ba2b1-c037-4949-94e4-ea9609405108/640/d27ba2b1-c037-4949-94e4-ea9609405108.png)
Synonym(s):
Erioglaucine disodium salt, Acid Blue 9, Alphazurine FG, FD&C;BLUE No. 1
Empirical Formula (Hill Notation):
C37H34Na2N2O9S3
CAS Number:
Molecular Weight:
792.85
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2