Skip to Content
Merck
Bisphenol A United States Pharmacopeia (USP) Reference Standard

Bisphenol A

Bisphenol A United States Pharmacopeia (USP) Reference Standard
Synonym(s):
2,2-Bis(4-hydroxyphenyl)propane, 4,4′-Isopropylidenediphenol
Linear Formula:
(CH3)2C(C6H4OH)2
CAS Number:
Molecular Weight:
228.29

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Monomer Type
Reference Material Type
Special Grade
Technique