Skip to Content
Merck
Bisoprolol for peak identification European Pharmacopoeia (EP) Reference Standard

Bisoprolol for peak identification

Bisoprolol for peak identification European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Bisoprolol hemifumarate salt, 1-[4-[[2-(1-Methylethoxy)ethoxy]methyl]phenoxy]-3-[(1-methylethyl)amino]-2-propanol hemifumarate salt
Empirical Formula (Hill Notation):
C18H31NO4 · .5C4H4O4
CAS Number:
Molecular Weight:
383.48

Products

Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1