Skip to Content
Merck
Bis(cyclopentadienyl)zirconium(IV) chloride hydride 95%

Bis(cyclopentadienyl)zirconium(IV) chloride hydride

Bis(cyclopentadienyl)zirconium(IV) chloride hydride 95%
Synonym(s):
Di(cyclopentadienyl)zirconium(IV) chloride hydride, Schwartz’ reagent, Zirconocene chloride hydride
Empirical Formula (Hill Notation):
C10H11ClZr
CAS Number:
Molecular Weight:
257.87
EC Number:

Products

Brand
Formula Weight
Physical Form
Purity
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing