
Bis(2,2,2-trifluoroethyl) ether

Synonym(s):
2,2,2-Trifluoroethyl ether, Flurothyl
Linear Formula:
(CF3CH2)2O
CAS Number:
Molecular Weight:
182.06
Products
Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1