![Benzophenone European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/185/054/3b3faf43-7dfe-4122-8581-77e1ceeabc4f/640/3b3faf43-7dfe-4122-8581-77e1ceeabc4f.png)
Benzophenone
![Benzophenone European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/185/054/3b3faf43-7dfe-4122-8581-77e1ceeabc4f/640/3b3faf43-7dfe-4122-8581-77e1ceeabc4f.png)
Synonym(s):
Benzophenone, Diphenyl ketone, Diphenylmethanone, NSC 8077
Linear Formula:
(C6H5)2CO
CAS Number:
Molecular Weight:
182.22
Products
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing