
Bendamustine Related Compound B

Synonym(s):
4-(1-Methyl-5-morpholino-1H-benzimidazol-2-yl)butanoic acid hydrochloride
Empirical Formula (Hill Notation):
C16H21N3O3xHCl
Products
Brand
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2