
β,β,3-Trimethylbenzenepropanol

Synonym(s):
2,2-Dimethyl-3-(3-methylphenyl)propanol, 2,2-Dimethyl-3-(3-tolyl)propan-1-ol, 3-(2,2-Dimethyl-3-hydroxypropyl)toluene
Empirical Formula (Hill Notation):
C12H18O
CAS Number:
Molecular Weight:
178.27
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing