![Bacillus spizizenii WDCM 00003 Vitroids™ 50,000-150,000 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/assets/images/pdp-no-image-sq/pdp-no-image-sq_w640.png)
Bacillus spizizenii WDCM 00003 Vitroids™
![Bacillus spizizenii WDCM 00003 Vitroids™ 50,000-150,000 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/assets/images/pdp-no-image-sq/pdp-no-image-sq_w640.png)
Synonym(s):
Bacillus subtilis subsp. spizizenii WDCM 00003 Vitroids™
Products
Brand
Manufacturer
Physical Form
Agency
Quality Segment
Application
Microorganism Suitability
Package Size
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing