![Azithromycin Related Compound F United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/176/575/1a302482-86d7-4275-a387-38467c666ada/640/1a302482-86d7-4275-a387-38467c666ada.png)
Azithromycin Related Compound F
![Azithromycin Related Compound F United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/176/575/1a302482-86d7-4275-a387-38467c666ada/640/1a302482-86d7-4275-a387-38467c666ada.png)
Synonym(s):
3′-N-Demethyl-3′-N-formylazithromycin
Empirical Formula (Hill Notation):
C38H70N2O13
CAS Number:
Molecular Weight:
762.97
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing