![Azinphos-methyl certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/428/463/5f6a98ec-86b6-489b-bf02-7299f5731192/640/5f6a98ec-86b6-489b-bf02-7299f5731192.png)
Azinphos-methyl
![Azinphos-methyl certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/428/463/5f6a98ec-86b6-489b-bf02-7299f5731192/640/5f6a98ec-86b6-489b-bf02-7299f5731192.png)
Synonym(s):
Guthion®, Methyltriazotin
Empirical Formula (Hill Notation):
C10H12N3O3PS2
CAS Number:
Molecular Weight:
317.32
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2