
Astemizole

Synonym(s):
1-(4-Fluorobenzyl)-2-(1-[4-methoxyphenethyl]piperidin-4-yl)aminobenzimidazole
Empirical Formula (Hill Notation):
C28H31FN4O
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing