Skip to Content
Merck
Ascorbic acid British Pharmacopoeia (BP) Reference Standard

Ascorbic Acid

Ascorbic acid British Pharmacopoeia (BP) Reference Standard
Synonym(s):
L-Ascorbic acid, L-Threoascorbic acid, Antiscorbutic factor, Vitamin C
Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
EC Number:

Products