Skip to Content
Merck
Ascentis® Express 90 Å PFAS, 2.7 μm HPLC Column column L × I.D. 25 cm × 2.1 mm, suitable for PFAS testing

Ascentis® Express 90 Å PFAS (2.7 μm) HPLC Columns

Ascentis® Express 90 Å PFAS, 2.7 μm HPLC Column column L × I.D. 25 cm × 2.1 mm, suitable for PFAS testing
Synonym(s):
Ascentis® Express 90 Å PFAS, 2.7 μm HPLC Column

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Application
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique