Skip to Content
Merck
Articaine Pharmaceutical Secondary Standard; Certified Reference Material

Articaine

Articaine Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Carticaine, Methyl 4-methyl-3-[2-(propylamino)propionamido]-2-thiophenecarboxylate, Articaine
Empirical Formula (Hill Notation):
C13H20N2O3S
CAS Number:
Molecular Weight:
284.37

Products

Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade