![Argatroban monohydrate](/deepweb/assets/sigmaaldrich/product/images/332/077/af09fdad-07a2-4363-a353-a33903ff75b8/640/af09fdad-07a2-4363-a353-a33903ff75b8.jpg)
Argatroban monohydrate
![Argatroban monohydrate](/deepweb/assets/sigmaaldrich/product/images/332/077/af09fdad-07a2-4363-a353-a33903ff75b8/640/af09fdad-07a2-4363-a353-a33903ff75b8.jpg)
Synonym(s):
(2R,4R)-1-[(2S)-5-[(Aminoiminomethyl)amino]-1-oxo-2-[[(1,2,3,4-tetrahydro-3-methyl-8-quinolinyl)sulfonyl]amino]pentyl]-4-methyl-2-piperidinecarboxylic Acid, Argipidine, MQPA
Empirical Formula (Hill Notation):
C23H36N6O5S · H2O
CAS Number:
Molecular Weight:
526.65
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Agency
Research Area
Quality Segment
Application
Package Size
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing