Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aquivion
®
E98-09S
Synonym(s):
Aquivion
®
SO
3
H
Linear Formula:
[CF
2
CF(OCF
2
CF
2
SO
3
H)]
m
[CF
2
CF
2
]
n
CAS Number:
1163733-25-2
Products
Brand
Sigma-Aldrich (1)
Physical Form
Length (cm)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
802735
Aquivion
®
E98-09S,
membrane sheet, contains CF
3
polymer chain ends as stabilizer, PFSA eq. wt. 980 g/mole SO
3
H, L × W × thickness 18 cm × 18 cm × 90 μm
Expand
View Pricing