Skip to Content
Merck
Anti-PRKAG2 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-PRKAG2 antibody produced in rabbit

Anti-PRKAG2 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-5′-AMP-activated protein kinase subunit γ-2 antibody produced in rabbit, Anti-AMPK γ-2 chain antibody produced in rabbit, Anti-AMPK γ2 antibody produced in rabbit, Anti-H91620p antibody produced in rabbit, 5′-AMP-activated protein kinase subunit γ-2, AAKG2, AMPK γ-2 chain, AMPK γ2, H91620p

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique