Anti-PPFIBP2 antibody produced in rabbit
Synonym(s):
Anti-Liprin-β-2 antibody produced in rabbit, Anti-PTPRF-interacting protein-binding protein 2 antibody produced in rabbit, Anti-Protein tyrosine phosphatase receptor type f polypeptide-interacting protein-binding protein 2 antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing