Skip to Content
Merck
Anti-Human IgE−Alkaline Phosphatase antibody, Mouse monoclonal clone GE-1, purified from hybridoma cell culture

Anti-Human IgE−Alkaline Phosphatase antibody, Mouse monoclonal

Anti-Human IgE−Alkaline Phosphatase antibody, Mouse monoclonal clone GE-1, purified from hybridoma cell culture
Synonym(s):
Monoclonal Anti-Human IgE

Products

Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique