![Anti-CLCC1 antibody produced in rabbit IgG fraction of antiserum](/deepweb/assets/sigmaaldrich/product/images/937/480/900932f8-da45-4b5f-a01d-3248dc630191/640/900932f8-da45-4b5f-a01d-3248dc630191.jpg)
Anti-CLCC1 antibody produced in rabbit
![Anti-CLCC1 antibody produced in rabbit IgG fraction of antiserum](/deepweb/assets/sigmaaldrich/product/images/937/480/900932f8-da45-4b5f-a01d-3248dc630191/640/900932f8-da45-4b5f-a01d-3248dc630191.jpg)
Synonym(s):
Anti-Mid-1-related chloride channel 1 isoform 1 antibody produced in rabbit, KIAA0761, MCLC, mid-1-related chloride channel 1, Anti-Chloride channel CLIC-like 1, Anti-KIAA0761, Anti-MCLC, Anti-RP11-475E11.6
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing