
Anti-CASC4 antibody produced in rabbit

Synonym(s):
Anti-DKFZp459F1927, Anti-H63, Anti-cancer susceptibility candidate 4
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing