
Anti-β-Actin (ACTB) Antibody

Synonym(s):
Monoclonal Anti-β-Actin, ACTB, Actin, cytoplasmic 1, Actin, cytoplasmic 1, N-terminally processed, Beta-actin, Anti-β-actin, Anti- PS1TP5BP1, Anti-BRWS1, Anti-actin, beta
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Isotype
Physical Form
Product Line
Recombinant Host
Antibody Form
Research Area
Conjugate
Quality Segment
Application
Post Translational Modification
Non-reactive Species
Species Reactivity
Technique
Showing 1-17 of 17
Compare
Product Number
Description
Pricing