![vibrant-m](/assets/images/pdp-no-image-sq/pdp-no-image-sq_w640.png)
Anastrozole Related Compound A
![vibrant-m](/assets/images/pdp-no-image-sq/pdp-no-image-sq_w640.png)
Synonym(s):
2,2′-(5-Methyl-1,3-phenylene)bis(2-methylpropanenitrile), α,α,α′,α′,5-Pentamethyl-1,3-benzenediacetonitrile, 3,5-Bis(2-cyano-2-methylethyl)toluene, 3,5-Bis(2-cyanoprop-2-yl)toluene, Anastrozole pentamethyl
Empirical Formula (Hill Notation):
C15H18N2
CAS Number:
Molecular Weight:
226.32
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2