![Amorolfine for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/315/896/0bfb42a3-5055-4c78-ad59-5003d3c467b6/640/0bfb42a3-5055-4c78-ad59-5003d3c467b6.png)
Amorolfine for system suitability
![Amorolfine for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/315/896/0bfb42a3-5055-4c78-ad59-5003d3c467b6/640/0bfb42a3-5055-4c78-ad59-5003d3c467b6.png)
Synonym(s):
Amorolfine hydrochloride, (2R,6S)-rel-4-[3-[4-(1,1-Dimethylpropyl)phenyl]-2-methylpropyl]-2,6-dimethyl-morpholine hydrochloride, Ro 14-4767/002
Empirical Formula (Hill Notation):
C21H35ON · HCl
CAS Number:
Molecular Weight:
353.97
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing