Ammonium-15N hydroxide solution
Linear Formula:
15NH4OH/H2O soln.
CAS Number:
Molecular Weight:
36.04
Products
Product Category
Brand
Formula Weight
Isotope
Markush Class
Markush Group
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing