Skip to Content
Merck
Amlexanox ≥98% (HPLC)

Amlexanox

Amlexanox ≥98% (HPLC)
Synonym(s):
2-Amino-7-(1-methylethyl)-5-oxo-5H-[1]Benzopyrano[2,3-b]pyridine-3-carboxylic acid, AA 673, Amoxanox, CHX 3673
Empirical Formula (Hill Notation):
C16H14N2O4
CAS Number:
Molecular Weight:
298.29

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing