Skip to Content
Merck
Aminoglutethimide European Pharmacopoeia (EP) Reference Standard

Aminoglutethimide

Aminoglutethimide European Pharmacopoeia (EP) Reference Standard
Synonym(s):
DL-Aminoglutethimide, 3-(4-Aminophenyl)-3-ethyl-2,6-piperidinedione, 3-(p-Aminophenyl)-3-ethylpiperidine-2,6-dione
Empirical Formula (Hill Notation):
C13H16N2O2
CAS Number:
Molecular Weight:
232.28

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade