Skip to Content
Merck
AM-2201 ≥98% (HPLC)

AM-2201

AM-2201 ≥98% (HPLC)
Synonym(s):
1-(5-Fluoropentyl)-3-(1-naphthoyl) indole, AM 2201, JWH 2201, [1-(5-Fluoropentyl)-1H-indol-3-yl]-1-naphthalenylmethanone
Empirical Formula (Hill Notation):
C24H22FNO
CAS Number:
Molecular Weight:
359.44

Products

Brand
Color
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing