Alprazolam Related Compound A
Synonym(s):
2-(2-Acetylhydrazino)-7-chloro-5-phenyl-3H-1,4-benzodiazepine
Empirical Formula (Hill Notation):
C17H15ClN4O
CAS Number:
Molecular Weight:
326.78
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2