Alprazolam
Synonym(s):
8-Chloro-1-methyl-6-phenyl-4H-[1,2,4]triazolo[4,3-a][1,4]benzodiazepine
Empirical Formula (Hill Notation):
C17H13ClN4
CAS Number:
Molecular Weight:
308.76
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing